Anti-KCNB1 Mouse Monoclonal Antibody [clone: S4-11]

Supplier: Rockland Immunochemicals
200-301-F97
RL200-301-F97EA 812.12 CAD
RL200-301-F97
Anti-KCNB1 Mouse Monoclonal Antibody [clone: S4-11]
Antibodies
Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S4-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR