Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Biotium

20352-1ML 20352-200UL
89411-124EA 535.73 CAD
89411-124 89411-538
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR