Anti-IgG Donkey Polyclonal Antibody (Biotin)

Supplier: Biotium

20191 20191-1
89138-776EA 411.57 CAD
89138-776 89138-778
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR