Anti-IgG Goat Polyclonal Antibody (CF™ 532)

Supplier: Biotium

20366-500UL 20366-50UL
89427-242EA 342.99 CAD
89427-242 89427-244
Anti-IgG Goat Polyclonal Antibody (CF™ 532)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 532
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR