Anti-IgG Rabbit Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20323 20323-1
89411-078EA 411.57 CAD
89411-078 89411-526
Anti-IgG Rabbit Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR