Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Supplier: Enzo Life Sciences
SPA-827D SPA-827F
CA200062-740EA 415.54 CAD
CA200062-740 CA200062-742
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Antibodies
Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mammals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR