Anti-IFN-g Mouse Monoclonal Antibody [clone: DB-1]

Supplier: Biolegend
507801 507802
CA507801-BLEA 161.18 CAD
CA507801-BL CA507802-BL
Anti-IFN-g Mouse Monoclonal Antibody [clone: DB-1]
Antibodies
Purified anti-rat IFN-γ [DB-1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ELISA Capture, ICFC, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, the suggested use of this reagent is 1-4 ug/ml. To obtain a linear standard curve, serial dilutions of IFN-γ recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate. The purified DB-1 has been tested by blocking fluorochrome conjugated DB-1 for intracellular cytokine staining. In order to obtain complete blocking results, a saturated amount of purified antibody ( 5.0 ug/million cells) should be used for incubation with target cells, prior to staining with fluorochrome conjugated antibody. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: DB-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR