Anti-Bcl-6 Mouse Monoclonal Antibody [clone: IG191E/A8]

Supplier: Biolegend
648301 648302
CA648301-BLEA 225.01 CAD
CA648301-BL CA648302-BL
Anti-Bcl-6 Mouse Monoclonal Antibody [clone: IG191E/A8]
Antibodies
Purified anti-mouse/human Bcl-6 [IG191E/A8]; Isotype: Mouse IgG1; Reactivity: Human, Mouse; Apps: WB, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 1.0 μg per ml (1:500). It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-6
Clonality: Monoclonal
Clone: IG191E/A8
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR