Anti-IL-12 p40 Rat Monoclonal Antibody [clone: C15.6]
Supplier: Biolegend
505201 505202
CA505201-BLEA 132.45 CAD
CA505201-BL CA505202-BL
Anti-IL-12 p40 Rat Monoclonal Antibody [clone: C15.6]
Antibodies
Purified anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Capture, IHC, IP, WB; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-12 recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performanceMore Product Information