Anti-FOXP3 Rat Monoclonal Antibody [clone: MF-14]

Supplier: Biolegend
126401 126402
CA126401-BLEA 205.86 CAD
CA126401-BL CA126402-BL
Anti-FOXP3 Rat Monoclonal Antibody [clone: MF-14]
Antibodies
Purified anti-mouse FOXP3 [MF-14]; Isotype: Rat IgG2b, κ; Reactivity: mouse; Apps: WB, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 1 μl/ml antibody dilution buffer for each mini-gel. Each lot of this antibody is quality control tested by immunofluorescent intracellular staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: MF-14
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR