Anti-CD146 Rat Monoclonal Antibody [clone: ME-9F1]

Supplier: Biolegend
134701 134702
CA134701-BLEA 161.18 CAD
CA134701-BL CA134702-BL
Anti-CD146 Rat Monoclonal Antibody [clone: ME-9F1]
Antibodies
Purified anti-mouse CD146 [ME-9F1]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: FC, IHC-F; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunohistochemistry, a concentration range of 5.0 - 10.0 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: ME-9F1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR