Anti-Notch 3 Mouse Monoclonal Antibody [clone: MHN3-21]

Supplier: Biolegend
345401 345402
CA345401-BLEA 178.73 CAD
CA345401-BL CA345402-BL
Anti-Notch 3 Mouse Monoclonal Antibody [clone: MHN3-21]
Antibodies
Purified anti-human Notch 3 [MHN3-21]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.03 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Notch 3
Clonality: Monoclonal
Clone: MHN3-21
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR