Anti-CCL2 Mouse Monoclonal Antibody [clone: 5D3-F7]

Supplier: Biolegend
502601 502602
CA502601-BLEA 150.01 CAD
CA502601-BL CA502602-BL
Anti-CCL2 Mouse Monoclonal Antibody [clone: 5D3-F7]
Antibodies
Purified anti-human MCP-1 [5D3-F7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: ELISA Capture, WB, ELISA, ELISPOT Detection, ICFC, IHC, IP; Size: 500 μg

Recommended Usage:  Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-8 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of MCP-1 recombinant protein ranging from 2000 to 31.3 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 5D3-F7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR