Anti-IL-13 Rat Monoclonal Antibody [clone: JES10-5A2]

Supplier: Biolegend
501901 501902
CA501901-BLEA 169.15 CAD
CA501901-BL CA501902-BL
Anti-IL-13 Rat Monoclonal Antibody [clone: JES10-5A2]
Antibodies
Purified anti-human IL-13 [JES10-5A2]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of human IL-13 recombinant protein ranging from 2000 to 8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-13
Clonality: Monoclonal
Clone: JES10-5A2
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR