Anti-FOXP3 Mouse Monoclonal Antibody [clone: 259D]

Supplier: Biolegend
320201 320202
CA320201-BLEA 205.86 CAD
CA320201-BL CA320202-BL
Anti-FOXP3 Mouse Monoclonal Antibody [clone: 259D]
Antibodies
Purified anti-human FOXP3 [259D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee; Apps: WB, IHC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent intracellular staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per 106 cells in 100 μl volume. For Western blotting, the suggested working dilution(s) is 5.0 μg/ml in antibody dilution buffer. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 259D
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR