Anti-DLL4 Mouse Monoclonal Antibody [clone: MHD4-46]

Supplier: Biolegend
346501 346502
CA346501-BLEA 113.3 CAD
CA346501-BL CA346502-BL
Anti-DLL4 Mouse Monoclonal Antibody [clone: MHD4-46]
Antibodies
Purified anti-human Delta-like protein 4 (DLL4) [MHD4-46]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: MHD4-46
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR