Anti-C3a/C3a(desArg)/C3 Mouse Monoclonal Antibody [clone: K13/16]

Supplier: Biolegend
518101 518102
CA518101-BLEA 161.18 CAD
CA518101-BL CA518102-BL
Anti-C3a/C3a(desArg)/C3 Mouse Monoclonal Antibody [clone: K13/16]
Antibodies
Purified anti-human C3a/C3a(desArg)/C3 [K13/16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of C3a recombinant protein ranging from 39 to 2500 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: C3a/C3a(desArg)/C3
Clonality: Monoclonal
Clone: K13/16
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR