Anti-Cyclin B1 Mouse Monoclonal Antibody [clone: V152]

Supplier: Biolegend
647901 647902
CA647901-BLEA 197.88 CAD
CA647901-BL CA647902-BL
Anti-Cyclin B1 Mouse Monoclonal Antibody [clone: V152]
Antibodies
Purified anti-Cyclin B1 [V152]; Isotype: Mouse IgG1; Reactivity: Human, Cross-Reactivity: Mouse; Apps: WB, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 2.5-5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Cyclin B1
Clonality: Monoclonal
Clone: V152
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human;Cross-Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR