Anti-CD199 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CW-1.2]

Supplier: Biolegend
128711 128712
CA128711-BLEA 197.88 CAD
CA128711-BL CA128712-BL
Anti-CD199 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CW-1.2]
Antibodies
PE/Cy7 anti-mouse CD199 (CCR9) [CW-1.2]; Isotype: Mouse IgG2a; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD199
Clonality: Monoclonal
Clone: CW-1.2
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR