Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 29-2L17]

Supplier: Biolegend
129815 129816
CA129815-BLEA 280.86 CAD
CA129815-BL CA129816-BL
Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 29-2L17]
Antibodies
PE/Cy7 anti-mouse CD196 (CCR6) [29-2L17]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD196 (CCR6)
Clonality: Monoclonal
Clone: 29-2L17
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR