Anti-IL-4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-81]

Supplier: Biolegend
511905 511906
CA511905-BLEA 194.69 CAD
CA511905-BL CA511906-BL
Anti-IL-4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-81]
Antibodies
PE anti-rat IL-4 [OX-81]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: OX-81
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR