Anti-CCL2 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H5]

Supplier: Biolegend
505903 505904
CA505903-BLEA 186.71 CAD
CA505903-BL CA505904-BL
Anti-CCL2 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H5]
Antibodies
PE anti-mouse/rat/human MCP-1 [2H5]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat, Human, Cross-Reactivity: Rhesus; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining by flow cytometry, the suggested use of this product is 0.5 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 2H5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat;Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR