Anti-Ly-6D Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49-H4]

Supplier: Biolegend
138603 138604
CA138603-BLEA 165.96 CAD
CA138603-BL CA138604-BL
Anti-Ly-6D Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49-H4]
Antibodies
PE anti-mouse Ly-6D [49-H4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6D
Clonality: Monoclonal
Clone: 49-H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR