Anti-IFN-g Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: XMG1.2]

Supplier: Biolegend
505807 505808
CA505807-BLEA 62.24 CAD
CA505807-BL CA505808-BL
Anti-IFN-g Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: XMG1.2]
Antibodies
PE anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR