Anti-Galectin-9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RG9-35]

Supplier: Biolegend
136103 136104
CA136103-BLEA 228.2 CAD
CA136103-BL CA136104-BL
Anti-Galectin-9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RG9-35]
Antibodies
PE anti-mouse Galectin-9 [RG9-35]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 200 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Galectin-9
Clonality: Monoclonal
Clone: RG9-35
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR