Anti-CXCL9 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MIG-2F5.5]

Supplier: Biolegend
515603 515604
CA515603-BLEA 341.5 CAD
CA515603-BL CA515604-BL
Anti-CXCL9 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MIG-2F5.5]
Antibodies
PE anti-mouse CXCL9 (MIG) [MIG-2F5.5]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CXCL9
Clonality: Monoclonal
Clone: MIG-2F5.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR