Anti-CD41 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MWReg30]

Supplier: Biolegend
133905 133906
CA133905-BLEA 114.9 CAD
CA133905-BL CA133906-BL
Anti-CD41 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MWReg30]
Antibodies
PE anti-mouse CD41 [MWReg30]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: MWReg30
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR