Anti-CD272 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8F4]

Supplier: Biolegend
134803 134804
CA134803-BLEA 213.84 CAD
CA134803-BL CA134804-BL
Anti-CD272 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8F4]
Antibodies
PE anti-mouse CD272 (BTLA) [8F4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD272
Clonality: Monoclonal
Clone: 8F4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR