Anti-CD26 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: H194-112]

Supplier: Biolegend
137803 137804
CA137803-BLEA 186.71 CAD
CA137803-BL CA137804-BL
Anti-CD26 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: H194-112]
Antibodies
PE anti-mouse CD26 (DPP-4) [H194-112]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD26
Clonality: Monoclonal
Clone: H194-112
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR