Anti-CD256 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: A3D8]

Supplier: Biolegend
136703 136704
CA136703-BLEA 205.86 CAD
CA136703-BL CA136704-BL
Anti-CD256 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: A3D8]
Antibodies
PE anti-mouse CD256 (APRIL) [A3D8]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: ICFC; Size: 200 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD256
Clonality: Monoclonal
Clone: A3D8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR