Anti-IL-12 p40 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: C11.5]

Supplier: Biolegend
501806 501807
CA501806-BLEA 165.96 CAD
CA501806-BL CA501807-BL
Anti-IL-12 p40 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: C11.5]
Antibodies
PE anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C11.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR