Anti-T-bet Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 4B10]

Supplier: Biolegend
644807 644808
CA644807-BLEA 280.86 CAD
CA644807-BL CA644808-BL
Anti-T-bet Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 4B10]
Antibodies
Pacific Blue™ anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our True-Nuclear™ Transcription Factor Staining Protocol. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in a volume of 100 μl. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: T-bet
Clonality: Monoclonal
Clone: 4B10
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR