Anti-Helios Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: 22F6]

Supplier: Biolegend
137210 137220
CA137210-BLEA 336.71 CAD
CA137210-BL CA137220-BL
Anti-Helios Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: 22F6]
Antibodies
Pacific Blue™ anti-mouse/human Helios [22F6]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol.
For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Helios
Clonality: Monoclonal
Clone: 22F6
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR