Anti-Ly-6G Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 1A8]

Supplier: Biolegend
127611 127612
CA127611-BLEA 167.56 CAD
CA127611-BL CA127612-BL
Anti-Ly-6G Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 1A8]
Antibodies
Pacific Blue™ anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR