Anti-IL-17A Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TC11-18H10.1]

Supplier: Biolegend
506917 506918
CA506917-BLEA 197.88 CAD
CA506917-BL CA506918-BL
Anti-IL-17A Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TC11-18H10.1]
Antibodies
Pacific Blue™ anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: TC11-18H10.1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR