Anti-IL-10 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: JES5-16E3]
Supplier: Biolegend
505019 505020
CA505019-BLEA 234.58 CAD
CA505019-BL CA505020-BL
Anti-IL-10 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: JES5-16E3]
Antibodies
Pacific Blue™ anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flowMore Product Information