Anti-H-2Kd Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: SF1-1.1]

Supplier: Biolegend
116615 116616
CA116615-BLEA 178.73 CAD
CA116615-BL CA116616-BL
Anti-H-2Kd Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: SF1-1.1]
Antibodies
Pacific Blue™ anti-mouse H-2Kd [SF1-1.1]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse d haplotype, Weakly cross-reacts with H-2k haplotype, Does not cross react with other haplotypes (e.g., b, j, p, q, s, v).; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kd
Clonality: Monoclonal
Clone: SF1-1.1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse (SJL) IgG2a, κ
Reactivity: Mouse d haplotype;Weakly cross-reacts with H-2k haplotype;Does not cross react with other haplotypes (e.g.;b;j;p;q;s;v).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR