Anti-H-2Kb Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: AF6-88.5]

Supplier: Biolegend
116513 116514
CA116513-BLEA 178.73 CAD
CA116513-BL CA116514-BL
Anti-H-2Kb Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: AF6-88.5]
Antibodies
Pacific Blue™ anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR