Anti-FOXP3 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: MF-14]

Supplier: Biolegend
126409 126410
CA126409-BLEA 300.01 CAD
CA126409-BL CA126410-BL
Anti-FOXP3 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: MF-14]
Antibodies
Pacific Blue™ anti-mouse FOXP3 [MF-14]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: MF-14
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR