Anti-CD80 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: 16-10A1]

Supplier: Biolegend
104723 104724
CA104723-BLEA 188.3 CAD
CA104723-BL CA104724-BL
Anti-CD80 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: 16-10A1]
Antibodies
Pacific Blue™ anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Cross-Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR