Anti-CD49b (pan-NK cells) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: DX5]

Supplier: Biolegend
108917 108918
CA108917-BLEA 178.73 CAD
CA108917-BL CA108918-BL
Anti-CD49b (pan-NK cells) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: DX5]
Antibodies
Pacific Blue™ anti-mouse CD49b (pan-NK cells) [DX5]; Isotype: Rat IgM, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49b (pan-NK cells)
Clonality: Monoclonal
Clone: DX5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD49b (pan-NK cells)
Environmentally Preferable
Antigen synonyms VLA-2 α chain|Integrin α2 chain|α2 integrin|DX5|ITGA2
Antibody type Primary
Clonality Monoclonal
Conjugation PB (Pacific Blue)
Clone DX5
Reactivity Mouse
Host Rat
Gene ID 16398
Isotype IgM kappa
Flow cytometry Yes
Immunogen IL-2-propagated NK1.1+ cells from C57BL/6 mice
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR