Anti-CD31 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 390]

Supplier: Biolegend
102421 102422
CA102421-BLEA 197.88 CAD
CA102421-BL CA102422-BL
Anti-CD31 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 390]
Antibodies
Pacific Blue™ anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 390
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR