Anti-CD105 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: MJ7/18]

Supplier: Biolegend
120411 120412
CA120411-BLEA 161.18 CAD
CA120411-BL CA120412-BL
Anti-CD105 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: MJ7/18]
Antibodies
Pacific Blue™ anti-mouse CD105 [MJ7/18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR