Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: C398.4A]

Supplier: Biolegend
313521 313522
CA313521-BLEA 169.15 CAD
CA313521-BL CA313522-BL
Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: C398.4A]
Antibodies
Pacific Blue™ anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: C398.4A
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Human;African Green;Baboon;Cynomolgus;Mouse;Rat;Rhesus;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR