Anti-CD49f Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GoH3]

Supplier: Biolegend
313619 313620
CA313619-BLEA 225.01 CAD
CA313619-BL CA313620-BL
Anti-CD49f Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GoH3]
Antibodies
Pacific Blue™ anti-human/mouse CD49f [GoH3]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Mouse, Baboon, Capuchin Monkey, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Rhesus, Sheep (Ovine), Swine (Pig, Porcine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49f
Clonality: Monoclonal
Clone: GoH3
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;African Green;Mouse;Baboon;Capuchin Monkey;Cat (Feline);Cattle (Bovine;Cow);Chimpanzee;Cynomolgus;Dog (Canine);Horse (Equine);Rabbit (Lapine);Rhesus;Sheep (Ovine);Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR