Anti-Granzyme A Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: CB9]

Supplier: Biolegend
507207 507208
CA507207-BLEA 422.89 CAD
CA507207-BL CA507208-BL
Anti-Granzyme A Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: CB9]
Antibodies
Pacific Blue™ anti-human Granzyme A [CB9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Granzyme A
Clonality: Monoclonal
Clone: CB9
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR