Anti-FOXP3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 259D]

Supplier: Biolegend
320215 320216
CA320215-BLEA 300.01 CAD
CA320215-BL CA320216-BL
Anti-FOXP3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 259D]
Antibodies
Pacific Blue™ anti-human FOXP3 [259D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 259D
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR