Anti-CD63 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: H5C6]

Supplier: Biolegend
353011 353012
CA353011-BLEA 225.01 CAD
CA353011-BL CA353012-BL
Anti-CD63 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: H5C6]
Antibodies
Pacific Blue™ anti-human CD63 [H5C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: H5C6
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR