Anti-CD5 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHT2]

Supplier: Biolegend
300623 300624
CA300623-BLEA 197.88 CAD
CA300623-BL CA300624-BL
Anti-CD5 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHT2]
Antibodies
Pacific Blue™ anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Capuchin Monkey;Common Marmoset;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR