Anti-CD335 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 9E2]

Supplier: Biolegend
331911 331912
CA331911-BLEA 225.01 CAD
CA331911-BL CA331912-BL
Anti-CD335 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 9E2]
Antibodies
Pacific Blue™ anti-human CD335 (NKp46) [9E2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or per 100 μl whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD335
Clonality: Monoclonal
Clone: 9E2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR